Anti-MSLN Rabbit Polyclonal Antibody

Supplier: ProSci Inc.
22-043
PRSI22-043EA 419 EUR
PRSI22-043
Anti-MSLN Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: MSLN
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR