Anti-UBE2J2 Rabbit Polyclonal Antibody

Supplier: ProSci Inc.
22-858
PRSI22-858EA 419 EUR
PRSI22-858
Anti-UBE2J2 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: UBE2J2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR