Anti-HBP1 Rabbit Polyclonal Antibody
Supplier: ProSci Inc.
25-467
PRSI25-467EA 660 EUR
PRSI25-467
Anti-HBP1 Rabbit Polyclonal Antibody
Antibodies
HBP1 is a transcriptional repressor that binds to the promoter region of target genes. This protein plays a role in the regulation of the cell cycle and of the Wnt pathway. It binds preferentially to the sequence 5'-TTCATTCATTCA-3'. Binding to the H1F0 promoter, HBP1 is enhanced by interaction with RB1. The protein also can disrupt the interaction between DNA and TCF4.
Antibody is purified by peptide affinity chromatography method.
Antibody is lyophilized in PBS buffer with 2% sucrose. Add 50 μL of distilled water. Final antibody concentration is 1 mg/mL.
Aliquot and store at -20°CMore Product Information