Anti-FZD9 Rabbit Polyclonal Antibody
Supplier: ProSci Inc.
25-681
PRSI25-681EA 660 EUR
PRSI25-681
Anti-FZD9 Rabbit Polyclonal Antibody
Antibodies
FZD9 contains 1 FZ (frizzled) domain and belongs to the G-protein coupled receptor Fz/Smo family. It is receptor for Wnt proteins. Most of frizzled receptors are coupled to the beta-catenin canonical signaling pathway, which leads to the activation of disheveled proteins, inhibition of GSK-3 kinase, nuclear accumulation of beta-catenin and activation of Wnt target genes. A second signaling pathway involving PKC and calcium fluxes has been seen for some family members. It may be involved in transduction and intercellular transmission of polarity information during tissue morphogenesis and/orMore Product Information