Anti-HAO2 Rabbit Polyclonal Antibody
Supplier: ProSci Inc.
25-700
PRSI25-700EA 660 EUR
PRSI25-700
Anti-HAO2 Rabbit Polyclonal Antibody
Antibodies
HAO2 is one of three related proteins that have 2-hydroxyacid oxidase activity yet differ in amino acid sequence, tissue expression and substrate preference. Subcellular location of the protein is the peroxisome. Specifically, the protein is expressed predominantly in liver and kidney and has the highest activity toward the substrate 2-hydroxypalmitate. Two alternatively spliced variants encoding the same isoform have been described.This gene is one of three related genes that have 2-hydroxyacid oxidase activity yet differ in encoded protein amino acid sequence, tissue expression andMore Product Information