Anti-ERMAP Rabbit Polyclonal Antibody

Supplier: ProSci Inc.
25-778
PRSI25-778EA 660 EUR
PRSI25-778
Anti-ERMAP Rabbit Polyclonal Antibody
Antibodies
ERMAP is a cell surface transmembrane protein that may act as an erythroid cell receptor, possibly as a mediator of cell adhesion. Polymorphisms in the gene encoding ERMAP protein are responsible for the Scianna/Radin blood group system. The protein encoded by this gene is a cell surface transmembrane protein that may act as an erythroid cell receptor, possibly as a mediator of cell adhesion. Polymorphisms in this gene are responsible for the Scianna/Radin blood group system. Two transcript variants encoding the same protein have been found for this gene.

Antibody is purified by peptideMore Product Information
Order Now

SPECIFICATIONS

Antigen symbol ERMAP
Antigen name Erythroblast membrane-associated Protein (Scianna blood group)
Environmentally Preferable
Antigen synonyms SC|RD|BTN5|PRO2801
Clonality Polyclonal
Conjugation Unconjugated
Reactivity Human,Rat,Mouse
Host Rabbit
Isotype
Western blot Yes
ELISA Yes
Immunogen A synthetic peptide corresponding a region of human ERMAP.
Form Lyophilized

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR