Anti-TSPAN17 Rabbit Polyclonal Antibody

Supplier: ProSci Inc.
25-811
PRSI25-811EA 660 EUR
PRSI25-811
Anti-TSPAN17 Rabbit Polyclonal Antibody
Antibodies
The function of TSPAN17 remains unknown.

Antibody is purified by peptide affinity chromatography method.

Antibody is lyophilized in PBS buffer with 2% sucrose. Add 50 μL of distilled water. Final antibody concentration is 1 mg/mL.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type:
Antigen: TSPAN17
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat
Order Now

SPECIFICATIONS

Antigen symbol TSPAN17
Antigen name Tetraspanin 17
Environmentally Preferable
Clonality Polyclonal
Conjugation Unconjugated
Reactivity Human,Rat,Mouse
Host Rabbit
Isotype
Western blot Yes
ELISA Yes
Immunogen A synthetic peptide corresponding a region of human TSPAN17.
Form Lyophilized

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR