Anti-SLC15A2 Rabbit Polyclonal Antibody

Supplier: ProSci Inc.
25-917
PRSI25-917EA 660 EUR
PRSI25-917
Anti-SLC15A2 Rabbit Polyclonal Antibody
Antibodies
SLC15A2 belongs to the PTR2/POT transporter (TC 2.A.17) family. It is a multi-pass membrane protein. The expression/activity of PEPT2 (SLC15A2) may be a critical factor in the modulation of opioidergic neurotransmission in vivo.

Antibody is purified by peptide affinity chromatography method.

Antibody is lyophilized in PBS buffer with 2% sucrose. Add 50 μL of distilled water. Final antibody concentration is 1 mg/mL.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type:
Antigen: SLC15A2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR