Anti-AQP10 Rabbit Polyclonal Antibody
Supplier: ProSci Inc.
26-497
PRSI26-497EA 660 EUR
PRSI26-497
Anti-AQP10 Rabbit Polyclonal Antibody
Antibodies
AQP10 is a member of the aquaglyceroporin family of integral membrane proteins. Members of this family function as water-permeable channels in the epithelia of organs that absorb and excrete water. AQP10 was shown to function as a water-selective channel, and could also permeate neutral solutes such as glycerol and urea.This gene encodes a member of the aquaglyceroporin family of integral membrane proteins. Members of this family function as water-permeable channels in the epithelia of organs that absorb and excrete water. This protein was shown to function as a water-selective channel, andMore Product Information