Anti-TFAP2E Rabbit Polyclonal Antibody

Supplier: ProSci Inc.
29-175
PRSI29-175EA 601 EUR
PRSI29-175
Anti-TFAP2E Rabbit Polyclonal Antibody
Antibodies
TFAP2E belongs to AP-2 family of transcription factors which play an important role in regulating gene expression during development and differentiation of multiple organs and tissues. It may play an important role in skin biology.

Antibody is purified by protein A chromatography method.

Antibody is lyophilized in PBS buffer with 2% sucrose. Add 50 μL of distilled water. Final antibody concentration is 1 mg/mL.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type:
Antigen: TFAP2E
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
More Product Information
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR