Anti-MPV17 Goat Polyclonal Antibody

Supplier: ProSci Inc.
42-674
PRSI42-674EA 620 EUR
PRSI42-674
Anti-MPV17 Goat Polyclonal Antibody
Antibodies
MPV17 antibody was purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide.

MPV17 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type:
Antigen: MPV17
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human
Order Now

SPECIFICATIONS

Antigen symbol MPV17
Environmentally Preferable
Antigen name MpV17 Mitochondrial Inner Membrane Protein
Antigen synonyms SYM1|MTDPS6
Clonality Polyclonal
Conjugation Unconjugated
Reactivity Human
Host Goat
Isotype
Western blot Yes
ELISA Yes
Immunogen MPV17 antibody was raised against a 12 amino acid synthetic peptide near the internal region of MPV17.
Form Liquid
Concentration 500 ug/mL

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR