Anti-ELF5 Goat Polyclonal Antibody

Supplier: ProSci Inc.
45-528
PRSI45-528EA 528 EUR
PRSI45-528
Anti-ELF5 Goat Polyclonal Antibody
Antibodies
ELF5 antibody was purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide.

ELF5 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type:
Antigen: ELF5
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human
Order Now

SPECIFICATIONS

Antigen symbol ELF5
Environmentally Preferable
Antigen name E74-like Factor 5 (ets Domain transcription Factor)
Antigen synonyms ESE-2|ETS-related transcription factor Elf-5|Epithelium-restricted ESE-1-related Ets factor|Epithelium-specific Ets transcription factor 2|ESE2|ELF5|E74-like factor 5
Clonality Polyclonal
Conjugation Unconjugated
Reactivity Human
Host Goat
Isotype
ELISA Yes
Immunogen ELF5 antibody was raised against a 12 amino acid synthetic peptide near the C-Terminus of ELF5.
Form Liquid
Concentration 500 ug/mL

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR