Anti-Tankyrase 1 Binding Protein 1 Goat Polyclonal Antibody

Supplier: ProSci Inc.
46-464
PRSI46-464EA 620 EUR
PRSI46-464
Anti-Tankyrase 1 Binding Protein 1 Goat Polyclonal Antibody
Antibodies
TAB182 antibody was purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide.

TAB182 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type:
Antigen: Tankyrase 1 Binding Protein 1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR