Anti-ENOX2 Goat Polyclonal Antibody

Supplier: ProSci Inc.
46-653
PRSI46-653EA 620 EUR
PRSI46-653
Anti-ENOX2 Goat Polyclonal Antibody
Antibodies
ELISA: Antibody detection limit dilution 1:64,000. Western Blot: Approximately 60 kDa and 37 kDa in human heart lysates after 0.03 μg/mL antibody staining.

ENOX2 antibody was purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide.

ENOX2 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type:
Antigen: ENOX2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human
Order Now

SPECIFICATIONS

Antigen symbol ENOX2
Environmentally Preferable
Clonality Polyclonal
Conjugation Unconjugated
Reactivity Human
Host Goat
Isotype
Western blot Yes
ELISA Yes
Immunogen ENOX2 antibody was raised against a 12 amino acid synthetic peptide near the internal region of ENOX2.
Form Liquid
Concentration 500 ug/mL

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR