Anti-GNPDA2 Rabbit Polyclonal Antibody

Supplier: ProSci Inc.
5061
PRSI5061EA 435 EUR
PRSI5061
Anti-GNPDA2 Rabbit Polyclonal Antibody
Antibodies
GNPDA2 Antibody: Glucosamine-6-phosphate deaminase (GNPDA) is an allosteric enzyme that catalyzes the reversible conversion of D-glucosamine-6-phosphate into D-fructose-6-phosphate and ammonium. GNPDA2 is the second mammalian glucosamine-6-phosphate deaminase discovered, and is closer in structure and activity to the E. coli enzyme than GNPDA1. GNPDA1 possesses greater affinity for ammonium than either GNPDA2 or the E. coli enzyme suggesting that the forward reaction of D-glucosamine-6-phosphate into D-fructose-6-phosphate and ammonium is catalyzed at a slower rate than GNPDA2.

Order Now

SPECIFICATIONS

Antigen symbol GNPDA2
Environmentally Preferable
Antigen name Glucosamine-6-phosphate Deaminase 2
Antigen synonyms SB52|GNPDA 2|Glucosamine-6-phosphate isomerase 2|Glucosamine-6-phosphate deaminase 2|GNP2
Clonality Polyclonal
Conjugation Unconjugated
Reactivity Human,Mouse
Host Rabbit
Isotype IgG
Western blot Yes
ELISA Yes
ImmunoChemistry Yes
ImmunoFluorescence Yes
Immunogen GNPDA2 antibody was raised against an 8 amino acid synthetic peptide near the carboxy terminus of human GNPDA2.
Purification Affinity chromatography purified via peptide column
Storage buffer PBS containing 0.02% sodium azide
Tested applications E, WB, ICC, IF

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR