Anti-LNPEP Rabbit Monoclonal Antibody [clone: E2]

Supplier: ProSci Inc.
54-325
PRSI54-325EA 654 EUR
PRSI54-325
Anti-LNPEP Rabbit Monoclonal Antibody [clone: E2]
Antibodies
Type: Primary
Antigen: LNPEP
Clonality: Monoclonal
Clone: E2
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR