Anti-GNMT Rabbit Polyclonal Antibody

Supplier: ProSci Inc.
59-615
PRSI59-615EA 620 EUR
PRSI59-615
Anti-GNMT Rabbit Polyclonal Antibody
Antibodies
For WB starting dilution is: 1:1000.

Type: Primary
Antigen: GNMT
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Amino Acid Range = 78-106
Host: Rabbit
Isotype: Ig
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR