Anti-ERBB3 Rabbit Polyclonal Antibody

Supplier: ProSci Inc.
63-071
PRSI63-071EA 620 EUR
PRSI63-071
Anti-ERBB3 Rabbit Polyclonal Antibody
Antibodies
For WB starting dilution is: 1:1000. For IHC-P starting dilution is: 1:10~50. For FACS starting dilution is: 1:10~50.

Type: Primary
Antigen: ERBB3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Amino Acid Range = 24-55
Host: Rabbit
Isotype: Ig
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR