Anti-ITGA6 Rabbit Polyclonal Antibody

Supplier: ProSci Inc.
63-858
PRSI63-858EA 620 EUR
PRSI63-858
Anti-ITGA6 Rabbit Polyclonal Antibody
Antibodies
For WB starting dilution is: 1:1000. For IHC-P starting dilution is: 1:50~100. For FACS starting dilution is: 1:10~50.

Type: Primary
Antigen: ITGA6
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Amino Acid Range = 1082-1110
Host: Rabbit
Isotype: Ig
Reactivity: Human
New Product
Order Now

SPECIFICATIONS

Antigen symbol ITGA6
Environmentally Preferable
Antigen name Integrin alpha-6
Antibody type Primary
Clonality Polyclonal
Conjugation Unconjugated
Reactivity Human
Host Rabbit
Gene ID 3655
Isotype
Western blot Yes
ImmunoChemistry Yes
Flow cytometry Yes
Modification None
Epitope 1082-1110 aa
Immunogen This CD49f antibody is generated from rabbits immunized with a KLH conjugated synthetic peptide between 1082-1110 amino acids from the C-terminal region of human CD49f.
Form Liquid
Molecular weight 127 kDa
Purification This antibody is prepared by Saturated Ammonium Sulfate (SAS) precipitation followed by dialysis
Concentration 2 mg/ml
Storage temperature Store at 4 °C for three months and –20 °C, stable for up to one year. As with all antibodies care should be taken to avoid repeated freeze thaw cycles. Antibodies should not be exposed to prolonged high temperatures.
Shipping temperature 4 °C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR