Anti-CD14 Mouse Monoclonal Antibody [clone: 61D3] (FITC (Fluorescein Isothiocyanate))

Supplier: ProSci Inc.
76-390
PRSI76-390EA 271 EUR
PRSI76-390
Anti-CD14 Mouse Monoclonal Antibody [clone: 61D3] (FITC (Fluorescein Isothiocyanate))
Antibodies
The 61D3 monoclonal antibody specifically reacts with CD14, a cell surface anchored glycoprotein that is primarily expressed on monocytes, inerfollicular macrophages, and a subset of dendritic cells. CD14 associates with MD-2 (LY-96) and TLR4 to form a receptor complex, which signals specifically in response to bacterial lipopolysaccharide (LPS) binding. The CD14/MD-2/TLR4 receptor complex signals via MyD88, TIRAP, and TRAF6, and ultimately activates NF-kappaB.

Type: Primary
Antigen: CD14
Clonality: Monoclonal
Clone: 61D3
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Mouse
More Product Information
New Product
Order Now

SPECIFICATIONS

Antigen symbol CD14
Antigen name CD14 molecule
Environmentally Preferable
Antigen synonyms CD14
Antibody type Primary
Clonality Monoclonal
Conjugation FITC (Fluorescein Isothiocyanate)
Clone 61D3
Reactivity
Host Mouse
Gene ID P08571
Isotype IgG1 kappa
Flow cytometry Yes
Form liquid
Purification The monoclonal antibody was purified utilizing affinity chromatography and unreacted dye was removed from the product.
Cross adsorption No
Storage buffer Phosphate-buffered aqueous solution, ≤0,09% Sodium azide, may contain carrier protein/stabiliser, ph 7,2.
Storage temperature The product should be stored undiluted at 4 °C and should be protected from prolonged exposure to light. Do not freeze.
Shipping temperature 4 °C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR