hPSC Genetic Analysis Kit
Supplier: STEMCELL Technologies
07550
STMC07550EA 1030 EUR
STMC07550
hPSC Genetic Analysis Kit
Nucleic Acid Reagents qPCR/RT-qPCR Enzymes and Kits
qPCR analysis kit for detecting the majority of karyotypic abnormalities reported in human ES and iPS cells.
Quickly and cost-effectively detect the 8 most common karyotypic abnormalities reported in human pluripotent stem cells (hPSCs).
This qPCR-based kit enables the genetic screening of multiple hPSC lines and contains enough material to analyse 20 individual samples in triplicate. It uses double-quenched probes with a 5-carboxyfluorescein (5-FAM) dye to give superior performance over other, single-quenched probes. The kit includes qPCR Master Mix, ROX Reference Dye, Buffer, and primer/probe mixes designed to detect the critical minimal regions of the 8 most commonly mutated regions, as well as a primer/probe mix to a control location in the genome. A genomic DNA control is also provided with demonstrated normal copy number over the regions of interest.
To understand your results, input qPCR data into our online Genetic Analysis App, which will perform statistical analyses, assist with data interpretation, and provide visual representation of the data.
Upozornení: Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').