EasySep™ Human Naïve B Cell Isolation Kits
Supplier: STEMCELL Technologies
17254RF 17254
STMC17254RFEA 2030 EUR
STMC17254RF STMC17254
EasySep™ Human Naïve B Cell Isolation Kits
Cell Separation Media
Immunomagnetic isolation of untouched human naïve B cells.
  • Fast, easy-to-use and column-free
  • Up to 98% purity with high recovery
  • Isolated cells are untouched
The EasySep™ Human Naïve B Cell Isolation Kit is designed to isolate naïve B cells from fresh or previously frozen peripheral blood mononuclear cells or washed leukapheresis samples by immunomagnetic negative selection. The EasySep™ procedure involves labeling unwanted cells with antibody complexes and magnetic particles. The magnetically labeled cells are separated from the untouched desiredMore Product Information