EasySep™ PE Positive Selection Kit II
Supplier: STEMCELL Technologies
17684 17684RF
STMC17684EA 1030 EUR
STMC17684 STMC17684RF
EasySep™ PE Positive Selection Kit II
Cell Separation Media
Immunomagnetic positive selection of any cells labeled with PE-conjugated antibodies.
  • Fast and easy-to-use
  • No columns required
The EasySep™ PE Positive Selection Kit II is designed to isolate cells that are labeled with PE-conjugated antibodies. Desired cells are targeted with antibody complexes recognising PE and dextran-coated magnetic particles. Labeled cells are separated using an EasySep™ magnet without the use of columns. Cells of interest remain in the tube while unwanted cells are poured off.
This product replaces the EasySep™ PE PositiveMore Product Information