EasySep™ Mouse CD19 Positive Selection Kit II
Supplier: STEMCELL Technologies
18954 18954RF
STMC18954EA 1490 EUR
STMC18954 STMC18954RF
EasySep™ Mouse CD19 Positive Selection Kit II
Cell Separation Media
Immunomagnetic positive selection of CD19+ cells (e.g. B cells) from mouse splenocytes or other tissues.
  • Fast and easy-to-use
  • Up to 99% purity
  • No columns required
  • Isolated cells are not fluorochrome-labelled
The EasySep™ Mouse CD19 Positive Selection Kit II is designed to isolate CD19+ cells from single-cell suspensions of splenocytes or other tissues by positive selection. Desired cells are targeted with antibody complexes recognising CD19 and dextran-coated magnetic particles. labelled cells are separated using an EasySep™ magnet without the use ofMore Product Information