EasySep™ Human Memory CD8+ T Cell Enrichment Kits
Supplier: STEMCELL Technologies
19159RF 19159
STMC19159RFEA 2000 EUR
STMC19159RF STMC19159
EasySep™ Human Memory CD8+ T Cell Enrichment Kits
Cell Separation Media
Immunomagnetic isolation of untouched human memory CD8+ T cells
  • Fast, easy-to-use and column-free
  • Up to 92% purity
  • Untouched, viable cells
The EasySep™ Human Memory CD8+ T Cell Enrichment Kit is designed to isolate memory CD8+ T cells from peripheral blood mononuclear cells by negative selection. Unwanted cells are targeted for removal with antibody complexes recognising CD4, CD14, CD16, CD19, CD20, CD34, CD36, CD45RA, CD56, CD61, CD66b, CD123, TCRγ/δ, glycophorin A and dextran-coated magnetic particles. The labelled cells are separated using anMore Product Information