EasySep™ Direct Human Monocyte Isolation Kits
Supplier: STEMCELL Technologies
19669 19669RF
STMC19669EA 1100 EUR
STMC19669 STMC19669RF
EasySep™ Direct Human Monocyte Isolation Kits
Cell Separation Media
Immunomagnetic isolation of monocytes directly from whole blood
  • 99,9% RBC depletion without the need for density gradient centrifugation, sedimentation, or lysis
  • Up to 92% purity of isolated cells
  • Fast, easy-to-use and column-free
  • Isolated cells are untouched
The EasySep™ Direct Human Monocyte Isolation Kit isolates functional, highly purified CD14+ monocytes directly from human whole blood by immunomagnetic negative selection. No lysis, density gradient centrifugation or other processing steps are required and isolated cells are immediately availableMore Product Information