Osteopontin, Corning®
Supplier: Corning
Corning®
BDAA354256EA 354.15 EUR
BDAA354256
Osteopontin, Corning®
Cell Culture Supplements and Additives
Osteopontin is found in a number of tissues, including bone, placenta, distal tubules of the kidney, and the central nervous system. Osteopontin is expressed in early bone development, at high levels at sites of bone remodelling, and has been implicated in the process of osteogenesis. Osteopontin is chemotactic for macrophages, smooth muscle cells, endothelial cells, and glial cells. In culture, osteopontin is used as an adhesive substrate for tumour cell lines and osteoclasts.