Phytohaemagglutinin-M (PHA-M)
Supplier: Biowest
L3010-005EA 0 EUR
L3010-005
Phytohaemagglutinin-M (PHA-M)
Cell Culture Supplements and Additives
Phytohaemagglutinin is a lectin extracted from red kidney beans (Phaseolus vulgaris). PHA-M is the mucoprotein form and is a crude extract used for the stimulation of cell proliferation in lymphocyte culture. PHA-M also has a powerful erythroagglutinating property and it was originally used for separating leukocytes from whole blood.