Biotin resin
Supplier: G-Biosciences
786-598EA 747 EUR
786-598
Biotin resin
Protein Extraction Protein Extraction Reagents
Immobilised biotin resin is designed for the high affinity chromatography purifications of avidin, streptavidin and neutravidin protein. The resin consists of biotin coupled to 6% cross-linked agarose.
  • Strong affinity for avidin, streptavidin and neutravidin
  • Reusable resin, at least 10 times
  • Covalently coupled to limit leaching