Anti-KIT Rat Monoclonal Antibody [clone: ACK2]

Supplier: Tonbo Biosciences
40-1172-U500 40-1172-U100
40-1172-U500EA 231 EUR
40-1172-U500 40-1172-U100
Anti-KIT Rat Monoclonal Antibody [clone: ACK2]
Antibodies
The ACK2 antibody is specific for CD117, also called c-Kit, a 145 kDa cytokine receptor important in the development of hematopoietic stem cells, in oogenesis, and for functional activity of immune cells such as NK and mast cells. c-Kit binds to a ligand known as stem cell factor (SCF), or alternatively as mast cell growth factor. Ligand binding promotes the activation (dimerization) and subsequent tyrosine kinase activity of the c-Kit receptor and triggers key survival, expansion and maturation signals during hematopoietic progenitor cell development. Conversely, shedding of extracellularMore Product Information
Order Now

SPECIFICATIONS

Antigen symbol KIT
Environmentally Preferable
Antigen name Tyrosine-protein kinase Kit
Antigen synonyms Fdc|SOW3|CD117|SCO1|Gsfsow3|W|Ssm|Tr-kit|SCO5|Gsfsco5|Gsfsco1|Bs|c-KIT
Antibody type Primary
Clonality Monoclonal
Conjugation Unconjugated
Clone ACK2
Reactivity Rat,Mouse
Host Rat
Gene ID 16590
Isotype IgG2b kappa
Western blot Yes
ImmunoChemistry Yes
Flow cytometry Yes
Cross adsorption none
Storage temperature Cool
Shipping temperature Ice

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR