Anti-CD45.2 Mouse Monoclonal Antibody (PE (Phycoerythrin)/Cy7®) [clone: 104]

Supplier: Tonbo Biosciences
60-0454-U025EA 41.6 EUR
60-0454-U025 60-0454-U100
Anti-CD45.2 Mouse Monoclonal Antibody (PE (Phycoerythrin)/Cy7®) [clone: 104]
Antibodies
The 104 antibody reacts with mouse CD45.2, also known as Ly5.2, which is a strain-specific allelic form of the CD45 Leukocyte Common Antigen (LCA). Functionally, CD45 is a protein tyrosine phosphatase whose broad cell distribution supports a critical role in many leukocyte functions, including regulation of signal transduction and cell activation associated with the T cell and B cell receptors. 

The 104 antibody is typically used as a leukocyte marker in Ly5.2 mouse strains C57BL/6, BALB/c, C58, DBA/1, DBA/2, C3H/He, CBA, 129, A and AKR. The antibody has been demonstrated to be specific for CD45.2 and is not cross-reactive with CD45.1-bearing cells.

Type: Primary
Antigen: CD45.2 (CD45.2)
Clonality: monoclonal
Clone: 104
Conjugation: PE (Phycoerythrin)/Cy7®
Epitope:
Host: Mouse
Isotype: IgG2a Kappa
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR