Anti-Ly-6G Rat Monoclonal Antibody [clone: RB6-8C5]

Supplier: Tonbo Biosciences
70-5931-U100 70-5931-U500
70-5931-U100EA 45.8 EUR
70-5931-U100 70-5931-U500
Anti-Ly-6G Rat Monoclonal Antibody [clone: RB6-8C5]
Antibodies
The RB6-8C5 antibody binds to mouse Ly-6G, commonly known as Gr-1, a member of the Ly-6 superfamily of GPI-anchored cell surface proteins with roles in cell signaling and cell adhesion. Gr-1 is differentially expressed during development and maturation of cells in the myeloid lineage and is expression at varying stages and levels on monocytes, macrophages, granulocytes, and peripheral neutrophils.

The RB6-8C5 antibody is typically used in combination with the macrophage labeling antibody M1/70 (Anti-CD11b) for phenotypic analysis of monocytes, macrophages and granulocytes. Note: The RB6-8More Product Information
Order Now

SPECIFICATIONS

Antigen symbol LY6C1
Antigen name Lymphocyte Antigen 6 Complex, locus C1
Environmentally Preferable
Antigen synonyms Ly6c|AA959465|Ly-6C|Ly-6C1|AA682074
Antibody type Primary
Clonality Monoclonal
Conjugation Unconjugated
Clone RB6-8C5
Reactivity Mouse
Host Rat
Gene ID 17067
Isotype IgG2b
ImmunoChemistry Yes
ImmunoPrecipitation Yes
Flow cytometry Yes
Neutralisation Yes
Cross adsorption none
Storage temperature Store undiluted at 2...8 °C, protect from light, do not freeze
Shipping temperature Ice
Tested applications Flow Cytometry

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR