Osteometric board
Supplier: Dynalon
DYLO8551-3224EA 0 EUR
DYLO8551-3224
Osteometric board
Educational Materials Forensics Educational Materials Crime Scene Evidence Learning Activities
Made of durable, lightweight polypropylene, this osteometric board is the most affordable on the market.
  • Easy to transport
  • Affordable
It features a sturdy fixed head piece and sliding foot piece that re operated with a smooth, fluid motion to allow for precise and accurate measurement of long bones up to 600 mm. The scale is accurate, easy-to-read and graduated in mm to eliminate the need for conversions in standard regression formulas.
L×W×H: 762×229×102 mm