Anti-IgY Goat Polyclonal Antibody (HRP (Horseradish Peroxidase))

Supplier: Tonbo Biosciences
72-8081-M001EA 140 EUR
72-8081-M001
Anti-IgY Goat Polyclonal Antibody (HRP (Horseradish Peroxidase))
Antibodies
Goat anti-Chicken IgY (H&L) - Affinity Pure, HRP Conjugate

Type: Secondary
Antigen: IgY
Clonality: Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope: Heavy and light chain
Host: Goat
Isotype:
Reactivity: Chicken
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR