J&W DB-608 GC columns
Supplier: Agilent
122-6832 123-1730 125-1730 125-6837
HEWL122-6832EA 815 EUR
HEWL122-6832 HEWL123-1730 HEWL125-1730 HEWL125-6837
J&W DB-608 GC columns
Chromatography Columns
DB-608 is specifically designed for the analysis of chlorinated pesticides and PCBs and is suitable for use in EPA methods 608, 508, and 8080.
  • Specifically designed for the analysis of chlorinated pesticides and PCBs
  • U.S. EPA Methods: 608, 508, 8080
  • Excellent inertness and recoveries without pesticide breakdown
  • Bonded and cross-linked
  • Solvent rinsable
J&W DB-608 columns are precision-engineered to deliver industry-leading performance in the effective analysis of chlorinated pesticides and PCBs. DB-608 widely used for U.S. EPA methods 608, 508, and 8080, and deliver excellent inertness and recoveries, without resulting in pesticide breakdown. The columns serve as exact replacements for HP-608 and are bonded, cross-linked, and solvent rinsable. DB-608 also feature operating temperature limits of 40 to 260/300 °C and are tested to some of the strictest industry QC standards to ensure the highest levels of quality every time.