Anti-F4/80 Rat Monoclonal Antibody [clone: BM8]

Supplier: STEMCELL Technologies
STMC60027EA 558 EUR
STMC60027
Anti-F4/80 Rat Monoclonal Antibody [clone: BM8]
Antibodies
Rat monoclonal IgG2a antibody against human, mouse F4/80, unconjugated.

  • Monocytes
  • Other

The BM8 antibody reacts with the F4/80 antigen (also termed Ly-71 in mouse and EMR-1 in human), an ~160 kDa transmembrane glycoprotein belonging to the EGF-TM7 family of G-protein-coupled receptors. F4/80 is considered a marker of choice for the identification of mature tissue macrophages, being broadly but variably expressed by this cell type in the liver (Kupffer cells), skin (Langerhans cells), bone marrow stroma, pancreas, thymus, spleen (red pulp), lung, and other tissues. It is also expressed by circulating monocytes, eosinophils and a subset of dendritic cells. F4/80 expression levels increase following activation of macrophages. F4/80 is reportedly the only macrophage marker suitable for distinguishing destructive from non-destructive inflammatory processes in the pancreas. The protein is thought to play a role in the generation of CD8+ regulatory T cells involved in immune tolerance. This antibody clone has been verified for purity assessments of cells isolated with EasySep™ kits, including EasySep™ Mouse Monocyte Enrichment Kit (Catalog #19761) and EasySep™ Mouse CD11b Positive Selection Kit (Catalog #18770).

Upozornení: Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').

Type:
Antigen: F4/80
Clonality: Monoclonal
Clone: BM8
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2a kappa
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR