Anti-Igg2a Isotype Control Mouse Monoclonal Antibody [clone: C1.18.4]

Supplier: STEMCELL Technologies
STMC60108EA 145 EUR
STMC60108
Anti-Igg2a Isotype Control Mouse Monoclonal Antibody [clone: C1.18.4]
Antibodies
Mouse monoclonal IgG2a, kappa isotype control antibody, unconjugated.

The C1.18.4 antibody (IgG2a, kappa) is suitable for use as an isotype-matched control antibody in several applications to estimate the degree of non-specific binding by an antigen-specific antibody. Ideally, the isotype control should have the same subclass of heavy chain (IgA, IgD, IgE, IgG, or IgM) and light chain (kappa or lambda) as the specific antibody being employed. If a conjugated antibody is employed, an isotype control conjugated to the same molecule (e.g. fluorochrome) should be chosen. The use of an appropriate isotype control helps confirm the specificity of the antigen-specific antibody and indicates non-specific binding that may result from binding to Fc receptors or other cell components. This antibody clone has been verified for use as an isotype control antibody for assessing non-specific binding to cells in applications such as flow cytometry.

Type:
Antigen: Igg2a Isotype Control
Clonality: Monoclonal
Clone: C1.18.4
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a kappa
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR