EasySep™ Multistand with EasySep™ Magnets
Supplier: STEMCELL Technologies
EasySep™
STMC18004EA 9860 EUR
STMC18004
EasySep™ Multistand with EasySep™ Magnets
Magnets
The EasySep™ Multistand allows the separation of up to four samples at one time.
  • Immunomagnetic column-free magnet
An ingenious reversible clamping bar allows the multistand to hold up to four EasySep™ magnets or four EasySep™ 'The Big Easy' magnets. After each round of magnetic separation, the supernatants from all samples are poured off simultaneously into receiving tubes. The receiving tube bar is indexed to ensure proper position of the tubes.
Balení: This set includes the EasySep™ multistand and four EasySep™ magnets.