EasySep™ Human CD11b Positive Selection and Depletion Kits
Supplier: STEMCELL Technologies
100-0742 100-0744
STMC100-0742EA 1120 EUR
STMC100-0742 STMC100-0744
EasySep™ Human CD11b Positive Selection and Depletion Kits
Cell Separation Media
Immunomagnetic positive selection of CD11b+ cells from human peripheral blood mononuclear cells (PBMCs) or leukapheresis samples.
  • Fast and easy-to-use
  • Up to 99% purity
  • No columns required
Efficiently isolate or deplete human CD11b+ cells from fresh or previously frozen peripheral blood mononuclear cells in as little as 16 minutes. Cells isolated with the EasySep™ Human CD11b Positive Selection and Depletion Kit can be used immediately in downstream applications such as flow cytometry, cell culture, or DNA/RNA extraction.
Use this kit to label CD11b+ cells with antibodies and magnetic particles. The cells are then subsequently separated—without the use of columns—using an EasySep™ magnet. Labeled cells remain in the tube while unlabeled cells are poured off. Depending on your research needs, either fraction can be retained for subsequent applications.
This kit targets human CD11b, a subunit of the integrin macrophage-1 antigen (Mac-1) receptor commonly used as a myeloid cell marker. CD11b is expressed on immune cells such as monocytes, macrophages, neutrophils, and NK cell subsets, and is known to play a role in regulating cell adhesion, migration, and phagocytosis
Upozornení: Product Restrictions: (i) Customer shall not make products or any portion of them, in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier; and (ii) Customer is familiar with Supplier’s restrictions on the use of the Products and shall use the Products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the ‘Product Restrictions’).