EasySep™ Non-Human Primate B Cell Isolation Kits
Supplier: STEMCELL Technologies
100-0345 100-0347
STMC100-0345EA 1970 EUR
STMC100-0345 STMC100-0347
EasySep™ Non-Human Primate B Cell Isolation Kits
Cell Separation Media
Immunomagnetic isolation of untouched non-human primate B cells.
  • Fast, easy to use, and column-free
  • Up to 95% purity
  • Untouched, viable cells
The EasySep™ Non-Human Primate B Cell Isolation Kit is designed to isolate untouched and highly purified B cells from fresh or previously frozen non-human primate peripheral blood mononuclear cells (PBMCs) in as little as 20 minutes by immunomagnetic negative selection. The EasySep™ procedure involves labeling unwanted cells with antibody complexes and magnetic particles. The magnetically labeled cells are separated from the untouched desired cells by using an EasySep™ magnet and simply pouring or pipetting the desired cells into a new tube. Isolated cells are immediately available for downstream applications such as flow cytometry, culture, or DNA/RNA extraction.
This kit has been verified for use with rhesus and cynomolgus macaques.
Upozornení: Product Restrictions: (i) Customer shall not make products or any portion of them, in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier; and (ii) Customer is familiar with Supplier’s restrictions on the use of the Products and shall use the Products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the ‘Product Restrictions’).