Bonded PP caps for sample storage vial kits
Supplier: Agilent
5183-4304 5183-4301 5183-4302 5183-4303 5183-4305 5183-4306 5183-4307 5183-4308
HEWL5183-4304EA 66.6 EUR
HEWL5183-4304 HEWL5183-4301 HEWL5183-4302 HEWL5183-4303 HEWL5183-4305 HEWL5183-4306 HEWL5183-4307 HEWL5183-4308
Bonded PP caps for sample storage vial kits
Closures Chromatography Vial Closures
Bonded caps include bonded septa that will prevent liner fallout during shipping and sampling. Caps are manufactured of high quality materials to eliminate sample contamination.